Thermo Scientific™
UltraFast™ UFC-200 GC Columns
 Related applications:
Industrial Chromatography
Dramatically increase the speed of your analysis with Thermo Scientific™ UltraFast™ UFC-200 GC Columns. If your laboratory processes large numbers of samples each day, UltraFast GC columns have the processing speed and throughput you need. These short, narrow-bore columns are designed for use with the Thermo Scientific™ TRACE GC™ UltraFast gas chromatograph configuration. Together they deliver a significant gain in analysis speed compared to conventional GC procedures.
Applications:
  • Chemical
  • Petrochemical
  • Environmental
  • Flavors and fragrances
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Tailored Specifically for Robustness and Speed
UltraFast GC columns utilize short (2–10m), narrow-bore capillaries and temperature programming conditions usually fast than 2°C/s, which results in peak widths in the 50-200ms range. As a result, analysis times average 1 minute or less. Benefits include:
  • Dramatically shorter analysis times
  • Increased sample throughput–by a factor of 20
  • Longer column lifetimes
UltraFast UFC-200 Columns Uses:
  • Alcohols
  • Ketones
Documents
Manuals & protocols