Search Thermo Fisher Scientific
Catalog Number | For Use With (Equipment) |
---|---|
WC34567 |
|
195964 | 13.0 cu. ft. -86°C Upright Freezers |
195965 | 17.3, 23, and 28 cu. ft. -86°C Upright Freezers |
195967 | 12.7, 17, and 20 cu. ft. -86°C Chest Freezers |
Work in a cooler environment with the Thermo Scientific™ Water-Cooled Condensers for -86°C upright and chest freezers, which emit 50% less heat into the lab compared to other freezers.
Requires:
Installation requires a qualified technician and should include proper adjustment of the regulating valve, which controls the discharge pressure.
Note:
Actual requirements for tower and city water sources may vary due to specific water pressure conditions.