Product Details

SNP ID
rs3092374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:49047407 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTCTGTTTCTTCCATATTCCAGG[C/G]TTATTCCGATTTAAAATTATTTGCA
Phenotype
MIM: 601342
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CSE1L PubMed Links

Gene Details

Gene
CSE1L
Gene Name
chromosome segregation 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256135.1 Intron NP_001243064.1
NM_001316.3 Intron NP_001307.2
XM_011528599.2 Intron XP_011526901.1
Gene
CSE1L-AS1
Gene Name
CSE1L antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details