Product Details
- SNP ID
-
hCV30837339
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:43371423 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCACCCCAGCCTTGGGTGTGCAGG[C/G]CTGAGATGGAGCATCATGATCACCC
- Phenotype
-
MIM: 613726
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ANO10
PubMed Links
Gene Details
- Gene
- ANO10
- Gene Name
- anoctamin 10
There are no transcripts associated with this gene.
View Full Product Details