The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008563.4 | NP_001008563.2 | 21 - 22 | 2613 | 73 |
NM_001110303.3 | NP_001103773.2 | 21 - 22 | 2515 | 73 | |
NM_006676.7 | NP_006667.3 | 21 - 22 | 2515 | 73 | |
XM_005251665.3 | XP_005251722.1 | 21 - 22 | 2503 | 73 | |
XM_005251666.3 | XP_005251723.1 | 16 - 17 | 2077 | 73 | |
XM_011518161.2 | XP_011516463.1 | 22 - 23 | 2855 | 73 | |
XM_011518162.2 | XP_011516464.1 | 21 - 22 | 3034 | 73 | |
XM_017014221.1 | XP_016869710.1 | 16 - 17 | 2077 | 73 | |
GenBank mRNA | AB023220.1 | - | 20 - 21 | 2433 | 73 |
AY074877.1 | - | 21 - 22 | 2549 | 73 | |
BC039593.1 | - | 21 - 22 | 2459 | 73 | |
BQ882312.1 | - | 4 - 5 | 530 | 73 |