Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC38A1
Entrez Gene ID
81539
Gene Name
solute carrier family 38 member 1
Gene Aliases
ATA1, NAT2, SAT1, SNAT1
Chromosome Location
Chr.12: 46183055 - 46269425 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077484.1 NP_001070952.1 14 - 15 1898 96
NM_001278387.1 NP_001265316.1 15 - 16 2360 96
NM_001278388.1 NP_001265317.1 13 - 14 2122 96
NM_001278389.1 NP_001265318.1 14 - 15 1387 96
NM_001278390.1 NP_001265319.1 14 - 15 2100 96
NM_030674.3 NP_109599.3 14 - 15 2237 96
XM_011538784.2 XP_011537086.1 14 - 15 1622 96
XM_011538786.2 XP_011537088.1 14 - 15 1622 96
XM_011538787.2 XP_011537089.1 14 - 15 1622 96
XM_017019990.1 XP_016875479.1 14 - 15 1852 96
GenBank mRNA AF271070.1 - 14 - 15 1481 96
AK027825.1 - 14 - 15 1387 96
AK074758.1 - 7 - 8 654 96
AK074797.1 - 11 - 12 956 96
AK074851.1 - 12 - 13 1099 96
AK074949.1 - 12 - 13 1145 96
AK225639.1 - 14 - 15 1475 96
AK225778.1 - 14 - 15 1497 96
AK309410.1 - 14 - 15 1814 96
AK313731.1 - 12 - 13 1188 96
BC010620.1 - 14 - 15 1444 96
BX647547.1 - 15 - 16 1932 96
BX648495.1 - 14 - 15 1825 96
BX648676.1 - 13 - 14 1692 96

View Full Product Details