Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KATNAL2
Entrez Gene ID
83473
Gene Name
katanin catalytic subunit A1 like 2
Gene Aliases
-
Chromosome Location
Chr.18: 46917602 - 47104795 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031303.2 NP_112593.2 14 - 15 1455 70
XM_005258357.4 XP_005258414.1 17 - 18 2000 70
XM_005258358.4 XP_005258415.1 17 - 18 1997 70
XM_005258361.3 XP_005258418.1 13 - 14 1257 70
XM_006722554.3 XP_006722617.1 17 - 18 2311 70
XM_017026028.1 XP_016881517.1 17 - 18 2026 70
XM_017026030.1 XP_016881519.1 16 - 17 1951 70
XM_017026031.1 XP_016881520.1 14 - 15 1436 70
GenBank mRNA BC034999.2 - 14 - 15 1455 70

View Full Product Details