Product Details

SNP ID
rs117224935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44317920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATTACAATACAGTGTCCCCAGCA[C/T]GGTGCCTGGCACAGGCAGGTGCTTA
Phenotype
MIM: 605448 MIM: 610820
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RUNDC3A PubMed Links

Gene Details

Gene
RUNDC3A
Gene Name
RUN domain containing 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144825.1 1829 Intron NP_001138297.1
NM_001144826.1 1829 Intron NP_001138298.1
NM_006695.4 1829 Intron NP_006686.1
XM_005256964.3 1829 Intron XP_005257021.1
XM_017024035.1 1829 Intron XP_016879524.1
XM_017024036.1 1829 Intron XP_016879525.1
XM_017024037.1 1829 Intron XP_016879526.1
XM_017024038.1 1829 UTR 3 XP_016879527.1
XM_017024039.1 1829 UTR 3 XP_016879528.1
XM_017024040.1 1829 UTR 3 XP_016879529.1
Gene
RUNDC3A-AS1
Gene Name
RUNDC3A antisense RNA 1
There are no transcripts associated with this gene.

Gene
SLC25A39
Gene Name
solute carrier family 25 member 39
There are no transcripts associated with this gene.

View Full Product Details