Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIGLEC11
Entrez Gene ID
102115985
Gene Name
sialic acid binding Ig like lectin 11
Gene Aliases
-
Chromosome Location
Chr.19: 50825096 - 50841016 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005596066.2 XP_005596123.2 4 - 5 933 64

View Full Product Details