Product Details

SNP ID
rs117080441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40031702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCCAGTTTTGTACTGAGTACAG[C/T]GTCAGCCATATCACACAAGTTGAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF780B PubMed Links

Gene Details

Gene
ZNF780B
Gene Name
zinc finger protein 780B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005851.2 5454 UTR 3 NP_001005851.1
XM_005258590.4 5454 UTR 3 XP_005258647.1
XM_005258591.4 5454 UTR 3 XP_005258648.1
XM_005258592.3 5454 UTR 3 XP_005258649.1
XM_005258593.2 5454 UTR 3 XP_005258650.1
XM_005258595.3 5454 UTR 3 XP_005258652.1
XM_006723072.3 5454 UTR 3 XP_006723135.1
XM_011526593.2 5454 UTR 3 XP_011524895.1
XM_017026425.1 5454 UTR 3 XP_016881914.1
XM_017026426.1 5454 UTR 3 XP_016881915.1
XM_017026427.1 5454 UTR 3 XP_016881916.1

View Full Product Details