Product Details
- SNP ID
-
rs201069805
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:75204246 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCGGGCGTGGTGGTAGAGCAGGTG[C/T]GGCTGCCCCAGGACACAATGCCCAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CTRB2
PubMed Links
Gene Details
- Gene
- CTRB2
- Gene Name
- chymotrypsinogen B2
View Full Product Details