Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rorb
Entrez Gene ID
309288
Gene Name
RAR-related orphan receptor B
Gene Aliases
-
Chromosome Location
Chr.1: 234252757 - 234442597 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270958.1 NP_001257887.1 7 - 8 1625 91
XM_002728869.3 XP_002728915.1 7 - 8 3277 91

View Full Product Details