Assay Design



Gene Transcripts

Gene Symbol
FABP7
Entrez Gene ID
777787
Gene Name
fatty acid binding protein 7
Gene Aliases
-
Chromosome Location
Chr.9: 28476061 - 28479847 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078162.2 NP_001071630.1 3 - 4 444 128
GenBank mRNA BC118369.1 - 3 - 4 445 128

View Full Product Details