Product Details

SNP ID
rs142215004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7796204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATGAAGGTAATGGTCCCATAGA[A/C]CAGGGTAACCACAGTGAGGTGGGAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR5P2 PubMed Links

Gene Details

Gene
OR5P2
Gene Name
olfactory receptor family 5 subfamily P member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153444.1 739 Missense Mutation GTC,TTC V247F NP_703145.1

View Full Product Details