Assay Design
Gene Transcripts
- Gene Symbol
- mas
- Entrez Gene ID
-
38499
- Gene Name
- masquerade
- Gene Aliases
-
CG15002, Dmel\CG15002, Dmel_CG15002, Mas, SPH79, c-SPH79, cSPH79
- Chromosome Location
-
Chr.3L: 4161666 - 4167464 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0011653
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079195.3 |
NP_523919.1 |
4 - 5 |
760 |
55 |
GenBank mRNA |
BT001597.1 |
- |
4 - 5 |
757 |
55 |
View Full Product Details