Gene Transcripts

Gene Symbol
Ccr4
Entrez Gene ID
171054
Gene Name
chemokine (C-C motif) receptor 4
Gene Aliases
Cmkbr4
Chromosome Location
Chr. 8: 122530178 - 122535959 on Build Rnor_6.0
UniGene ID
Rn.81076
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133532.2 NP_598216.2 1 248
XM_006243915.2 XP_006243977.1 2 435
GenBank mRNA AF432872.2 - 1 248

View Full Product Details