Gene Transcripts

Gene Symbol
Pdhb
Entrez Gene ID
68263
Gene Name
pyruvate dehydrogenase (lipoamide) beta
Gene Aliases
2610103L06Rik, AL024199, C81408
Chromosome Location
Chr. 14: 8165991 - 8172992 on Build GRCm38
UniGene ID
Mm.301527
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024221.3 NP_077183.1 4 282
GenBank mRNA AK011810.1 - 246
AK014180.1 - 4 238
AK014918.1 - 4 283
AK084507.1 - 3 1473
AK153058.1 - 4 236
AK163596.1 - 1 1652
AK166631.1 - 4 284
AK167798.1 - 4 239
BC002188.1 - 2 106
BC019512.1 - 4 255 4210204
BC094468.1 - 4 228 5011266
CJ159324.1 - 4 286

View Full Product Details