Product Details

SNP ID
rs141074846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132873957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCTTTGCCCTTACAGATAGATG[A/G]CTCCCCAATACCAGAAGTGAGTTCG
Phenotype
MIM: 604417 MIM: 601918 MIM: 611373 MIM: 612080
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AFF4 PubMed Links

Gene Details

Gene
AFF4
Gene Name
AF4/FMR2 family member 4
There are no transcripts associated with this gene.

Gene
GDF9
Gene Name
growth differentiation factor 9
There are no transcripts associated with this gene.

Gene
LEAP2
Gene Name
liver enriched antimicrobial peptide 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052971.2 94 Missense Mutation GAC,GGC D22G NP_443203.1
Gene
UQCRQ
Gene Name
ubiquinol-cytochrome c reductase complex III subunit VII
There are no transcripts associated with this gene.

View Full Product Details