Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kqt-1
Entrez Gene ID
180933
Gene Name
protein-coding
Gene Aliases
CELE_C25B8.1
Chromosome Location
Chr.X: 6619012 - 6627995 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171709.6 NP_741819.1 13 - 14 1509 156
NM_171710.7 NP_741820.1 12 - 13 1524 156
GenBank mRNA AY572974.1 - 12 - 13 1355 156

View Full Product Details