Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rsks-1
Entrez Gene ID
176554
Gene Name
Ribosomal protein S6 kinase beta
Gene Aliases
CELE_Y47D3A.16
Chromosome Location
Chr.III: 11245120 - 11252281 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067046.4 NP_499447.2 4 - 5 1516 61

View Full Product Details