Gene Transcripts
- Gene Symbol
- UNK
- Entrez Gene ID
- 85451
- Gene Name
- unkempt family zinc finger
- Gene Aliases
- UNKEMPT, ZC3H5, ZC3HDC5
- Chromosome Location
- Chr. 17: 75784771 - 75825805 on Build GRCh38
- UniGene ID
- Hs.584768
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001080419.2 |
NP_001073888.2
|
8, 9 |
1142 |
|
XM_006722145.3 |
XP_006722208.1
|
11, 12 |
5523 |
|
XM_006722148.2 |
XP_006722211.1
|
9, 10 |
1283 |
|
XM_011525400.1 |
XP_011523702.1
|
9, 10 |
1211 |
|
XM_011525401.2 |
XP_011523703.1
|
3, 4 |
299 |
|
XM_017025247.1 |
XP_016880736.1
|
11, 12 |
5523 |
|
XM_017025248.1 |
XP_016880737.1
|
10, 11 |
5425 |
|
XM_017025249.1 |
XP_016880738.1
|
10, 11 |
1568 |
|
XM_017025250.1 |
XP_016880739.1
|
9, 10 |
1470 |
|
GenBank mRNA |
AB051540.1 |
- |
8, 9 |
1125 |
|
BC053362.1 |
- |
8, 9 |
1125 |
|
View Full Product Details