The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080512.2 | NP_001073981.1 | 1 - 2 | 194 | 60 |
GenBank mRNA | CF242850.1 | - | 1 - 2 | 74 | 60 |