Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS20
Entrez Gene ID
80070
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 20
Gene Aliases
ADAM-TS20, ADAMTS-20, GON-1
Chromosome Location
Chr.12: 43352843 - 43551971 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025003.3 NP_079279.3 10 - 11 1505 88
GenBank mRNA AF488804.1 - 10 - 11 1505 88
AJ515153.3 - 10 - 11 1505 88
AJ515154.2 - 10 - 11 1505 88

View Full Product Details