Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cog1
Entrez Gene ID
16834
Gene Name
component of oligomeric golgi complex 1
Gene Aliases
6030445I15, Ldlb, mKIAA1381
Chromosome Location
Chr.11: 113649178 - 113669331 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013581.3 NP_038609.3 9 - 10 2409 63
GenBank mRNA AF109377.2 - 9 - 10 2392 63
AK031510.1 - 8 - 9 1827 63
AK034636.2 - 9 - 10 2414 63
AK152464.1 - 9 - 10 2400 63
AK158507.1 - 9 - 10 2771 63
AK159497.1 - 9 - 10 2411 63
AK171574.1 - 9 - 10 2403 63
AK177622.1 - 1 - 2 178 63
AK187486.1 - 1 - 2 178 63
AK189116.1 - 1 - 2 178 63
AK220387.1 - 8 - 9 2853 63
BC063056.1 - 9 - 10 2391 63

View Full Product Details