Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DHRS7
Entrez Gene ID
100152785
Gene Name
dehydrogenase/reductase 7
Gene Aliases
-
Chromosome Location
Chr.1: 189153579 - 189170745 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244160.1 NP_001231089.1 5 - 6 1100 95
GenBank mRNA AK231851.1 - 5 - 6 813 95
AK232729.1 - 5 - 6 872 95
AK239997.1 - 5 - 6 854 95
AK398059.1 - 5 - 6 866 95
AY609687.1 - 5 - 6 1100 95

View Full Product Details