Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIP2;3
Entrez Gene ID
100233094
Gene Name
aquaporin-like
Gene Aliases
VITISV_029527
Chromosome Location
Chr.8: 11871846 - 11873426 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA DQ834700.1 - 1 - 2 410 92
EF364437.1 - 1 - 2 385 92

View Full Product Details