Gene Transcripts

Gene Symbol
HSPB8
Entrez Gene ID
26353
Gene Name
heat shock protein family B (small) member 8
Gene Aliases
CMT2L, DHMN2, E2IG1, H11, HMN2, HMN2A, HSP22
Chromosome Location
Chr. 12: 119178790 - 119194746 on Build GRCh38
UniGene ID
Hs.400095
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014365.2 NP_055180.1 3 1041
GenBank mRNA AF133207.1 - 3 878
AF191017.1 - 3 1041
AF217987.1 - 3 886
AF250138.1 - 3 526
AK312501.1 - 3 894
AL136936.1 - 3 871
BC002673.2 - 3 1009 3607117
BT006876.1 - 3 518
CR533453.1 - 3 518

View Full Product Details