Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCR20
Entrez Gene ID
3771457
Gene Name
low-molecular-weight cysteine-rich 20
Gene Aliases
AT5G47075, low-molecular-weight cysteine-rich 20
Chromosome Location
Chr.5: 19122503 - 19122866 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036947.2 NP_001032024.1 2 - 2 178 107

View Full Product Details