Product Details

SNP ID
rs117307819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7963558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCGGTAGCCGTTCAGGCTGGC[A/T]ATGGCCATCGCGGCTTCTTCATAGT
Phenotype
MIM: 603466
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ELAVL1 PubMed Links

Gene Details

Gene
ELAVL1
Gene Name
ELAV like RNA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001419.2 1073 Silent Mutation ATA,ATT I302I NP_001410.2

View Full Product Details