Product Details
- SNP ID
-
rs12792886
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:34107174 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGCCTCTCCTGCAGCTCCACTAC[C/T]ACTCCCGGCTAAGTTTTGTATTTTT
- Phenotype
-
MIM: 601178
MIM: 609221
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CAPRIN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117850996] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CAPRIN1
- Gene Name
- cell cycle associated protein 1
There are no transcripts associated with this gene.
- Gene
- NAT10
- Gene Name
- N-acetyltransferase 10
View Full Product Details