Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92121762-92122302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF2
Entrez Gene ID
53981
Gene Name
cleavage and polyadenylation specific factor 2
Gene Aliases
CPSF100
UniGene
Hs.657632
Gene Chromosome Location
Chr.14: 92121954-92164199 on Build GRCh38
Transcripts
NM_001322270.1 NM_001322271.1 NM_001322272.1 NM_017437.2 NM_004545.3

Gene Symbol
NDUFB1
Entrez Gene ID
4707
Gene Name
NADH:ubiquinone oxidoreductase subunit B1
Gene Aliases
CI-MNLL, CI-SGDH, MNLL
UniGene
Hs.183435
Gene Chromosome Location
Chr.14: 92116123-92121809 on Build GRCh38
Transcripts
NM_001322270.1 NM_001322271.1 NM_001322272.1 NM_017437.2 NM_004545.3

SNP Detail

SNP ID Chromosome Location
rs180896870 Chr.14: 92122153-92122153 on Build GRCh38
rs921433300 Chr.14: 92121804-92121804 on Build GRCh38
rs895820304 Chr.14: 92121990-92121990 on Build GRCh38
rs931503980 Chr.14: 92121912-92121912 on Build GRCh38
rs185351986 Chr.14: 92122234-92122234 on Build GRCh38
rs917325322 Chr.14: 92122103-92122103 on Build GRCh38
rs200764044 Chr.14: 92121825-92121825 on Build GRCh38
rs71430749 Chr.14: 92122116-92122116 on Build GRCh38
rs767708489 Chr.14: 92122091-92122091 on Build GRCh38
rs1020648711 Chr.14: 92121998-92121998 on Build GRCh38
rs946071272 Chr.14: 92122108-92122108 on Build GRCh38
rs1004237410 Chr.14: 92122029-92122029 on Build GRCh38
rs1044169276 Chr.14: 92122210-92122210 on Build GRCh38
rs907036988 Chr.14: 92122003-92122003 on Build GRCh38
rs796251496 Chr.14: 92121812-92121815 on Build GRCh38
rs10603293 Chr.14: 92121822-92121825 on Build GRCh38
rs558781305 Chr.14: 92122250-92122250 on Build GRCh38
rs924897478 Chr.14: 92122099-92122099 on Build GRCh38
rs769846570 Chr.14: 92121885-92121885 on Build GRCh38
rs12886909 Chr.14: 92121848-92121848 on Build GRCh38
rs527978762 Chr.14: 92121955-92121955 on Build GRCh38
rs918828127 Chr.14: 92121895-92121895 on Build GRCh38
rs1047576958 Chr.14: 92122245-92122245 on Build GRCh38
rs77548492 Chr.14: 92121821-92121825 on Build GRCh38
rs975530482 Chr.14: 92122051-92122051 on Build GRCh38
rs1041714932 Chr.14: 92122128-92122128 on Build GRCh38
rs7148709 Chr.14: 92121875-92121875 on Build GRCh38
rs1000066590 Chr.14: 92122001-92122001 on Build GRCh38
rs547428304 Chr.14: 92121972-92121972 on Build GRCh38
rs1016152552 Chr.14: 92122265-92122265 on Build GRCh38
rs1008717381 Chr.14: 92122232-92122232 on Build GRCh38
rs1054036906 Chr.14: 92121963-92121963 on Build GRCh38
rs34361571 Chr.14: 92122265-92122266 on Build GRCh38
rs779186117 Chr.14: 92121830-92121831 on Build GRCh38
rs749518220 Chr.14: 92121812-92121813 on Build GRCh38
rs924845117 Chr.14: 92121926-92121926 on Build GRCh38
rs748135141 Chr.14: 92121789-92121789 on Build GRCh38
rs76987927 Chr.14: 92121824-92121827 on Build GRCh38
rs948356135 Chr.14: 92122186-92122186 on Build GRCh38
rs569760200 Chr.14: 92122187-92122187 on Build GRCh38
rs772822974 Chr.14: 92121798-92121798 on Build GRCh38
rs765622861 Chr.14: 92121810-92121810 on Build GRCh38
rs79132362 Chr.14: 92121861-92121861 on Build GRCh38
rs1028831415 Chr.14: 92122002-92122003 on Build GRCh38
rs866168810 Chr.14: 92122064-92122064 on Build GRCh38
rs776282140 Chr.14: 92121805-92121805 on Build GRCh38
rs953156893 Chr.14: 92122012-92122012 on Build GRCh38
rs141166703 Chr.14: 92121922-92121922 on Build GRCh38
rs756494972 Chr.14: 92121811-92121812 on Build GRCh38
rs771878698 Chr.14: 92121794-92121794 on Build GRCh38
rs778057599 Chr.14: 92121812-92121812 on Build GRCh38
rs886393649 Chr.14: 92122255-92122255 on Build GRCh38
rs1026248806 Chr.14: 92122010-92122010 on Build GRCh38
rs963152054 Chr.14: 92122037-92122037 on Build GRCh38
rs914231364 Chr.14: 92122174-92122174 on Build GRCh38
rs567636739 Chr.14: 92121993-92121993 on Build GRCh38
rs961814117 Chr.14: 92122241-92122241 on Build GRCh38
rs921310203 Chr.14: 92122053-92122053 on Build GRCh38
rs565179334 Chr.14: 92121812-92121816 on Build GRCh38
rs541576452 Chr.14: 92122042-92122042 on Build GRCh38
rs918869672 Chr.14: 92122020-92122020 on Build GRCh38
rs906917127 Chr.14: 92122220-92122220 on Build GRCh38
rs531902020 Chr.14: 92121880-92121880 on Build GRCh38
rs553728652 Chr.14: 92121816-92121816 on Build GRCh38
rs955587548 Chr.14: 92122060-92122060 on Build GRCh38
rs962767196 Chr.14: 92121790-92121790 on Build GRCh38
rs577930881 Chr.14: 92121815-92121816 on Build GRCh38
rs927476179 Chr.14: 92122061-92122061 on Build GRCh38
rs7148238 Chr.14: 92121803-92121803 on Build GRCh38
rs567838672 Chr.14: 92121814-92121816 on Build GRCh38
rs1054089781 Chr.14: 92122160-92122160 on Build GRCh38
rs1003094409 Chr.14: 92122006-92122006 on Build GRCh38
rs746501011 Chr.14: 92121802-92121802 on Build GRCh38
rs545307969 Chr.14: 92121883-92121883 on Build GRCh38
rs1016940504 Chr.14: 92122033-92122033 on Build GRCh38
rs554808665 Chr.14: 92121816-92121817 on Build GRCh38
rs1012666576 Chr.14: 92121992-92121992 on Build GRCh38
rs1046027510 Chr.14: 92122223-92122223 on Build GRCh38
rs1018672987 Chr.14: 92122017-92122017 on Build GRCh38
rs997231571 Chr.14: 92122275-92122275 on Build GRCh38
rs971743967 Chr.14: 92122018-92122018 on Build GRCh38
rs377341616 Chr.14: 92121787-92121787 on Build GRCh38
rs888970478 Chr.14: 92122229-92122229 on Build GRCh38
rs1006132634 Chr.14: 92122263-92122263 on Build GRCh38
rs565756487 Chr.14: 92121813-92121816 on Build GRCh38
rs950599676 Chr.14: 92122016-92122016 on Build GRCh38
rs564429144 Chr.14: 92122266-92122266 on Build GRCh38
rs904430701 Chr.14: 92122000-92122000 on Build GRCh38
rs757545489 Chr.14: 92121812-92121814 on Build GRCh38
rs993609994 Chr.14: 92122244-92122244 on Build GRCh38
rs17127959 Chr.14: 92122070-92122070 on Build GRCh38
rs759122951 Chr.14: 92121806-92121806 on Build GRCh38
rs764502742 Chr.14: 92121828-92121828 on Build GRCh38
rs1057112724 Chr.14: 92121980-92121980 on Build GRCh38

View Full Product Details