Product Details

SNP ID
rs13482015
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:111805643 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TATTTCTCTTCCAGAATCCTATTTA[A/G]TAAGGTTAATCTTTTAAAAATACAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Gm15327 PubMed Links

Gene Details

Gene
Gm15327
Gene Name
predicted gene 15327
There are no transcripts associated with this gene.

Gene
Map3k1
Gene Name
mitogen-activated protein kinase kinase kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_011945.2 Intron NP_036075.2

View Full Product Details