The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026582.4 | NP_080858.3 | 2 - 3 | 624 | 83 |
NR_037590.1 | 3 - 4 | 705 | 83 | ||
GenBank mRNA | AK019886.1 | - | 3 - 4 | 663 | 83 |
AK029053.1 | - | 2 - 3 | 583 | 83 | |
AK029104.1 | - | 2 - 3 | 581 | 83 | |
AK029315.1 | - | 2 - 3 | 581 | 83 | |
AK031444.1 | - | 2 - 3 | 581 | 83 | |
AK044810.1 | - | 2 - 3 | 583 | 83 | |
AK049842.1 | - | 2 - 3 | 581 | 83 | |
AK077407.1 | - | 2 - 3 | 582 | 83 | |
AK128912.1 | - | 2 - 3 | 580 | 83 | |
AK159207.1 | - | 2 - 3 | 581 | 83 | |
AK159977.1 | - | 2 - 3 | 583 | 83 | |
AK184430.1 | - | 2 - 3 | 324 | 83 | |
AK191077.1 | - | 2 - 3 | 83 | ||
AK213342.1 | - | 1 - 2 | 166 | 83 | |
AK214024.1 | - | 2 - 3 | 291 | 83 | |
BC018381.1 | - | 1 - 2 | 208 | 83 | |
BC075615.1 | - | 2 - 3 | 624 | 83 |