Gene Transcripts

Gene Symbol
Vmn2r78
Entrez Gene ID
637896
Gene Name
vomeronasal 2, receptor 78
Gene Aliases
Chromosome Location
Chr. 7: 86915349 - 86955177 on Build GRCm38
UniGene ID
Mm.389957
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105189.1 NP_001098659.1 4 1473

View Full Product Details