Gene Transcripts

Gene Symbol
SLC25A32
Entrez Gene ID
81034
Gene Name
solute carrier family 25 member 32
Gene Aliases
MFT, MFTC, RREI
Chromosome Location
Chr. 8: 103398638 - 103415335 on Build GRCh38
UniGene ID
Hs.532265
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030780.4 NP_110407.2 5 1039
NR_102337.1 4 953
NR_102338.1 6 1148
XM_017013877.1 XP_016869366.1 5 942
GenBank mRNA AF283645.1 - 5 769
AK027531.1 - 5 808
AK027787.1 - 5 773
AK125492.1 - 6 757
AK300748.1 - 4 687
BC021893.1 - 5 747 4524907
DC331449.1 - 4 451

View Full Product Details