Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027030.2 | NP_081306.1 | 3 | 434 | |
GenBank mRNA | AK005584.1 | - | 3 | 435 | |
AK169641.1 | - | 3 | 442 | ||
AK171018.1 | - | 3 | 426 | ||
AY040775.1 | - | 3 | 447 | ||
BC016273.1 | - | 3 | 454 | 4511427 |