Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 78867722-78868245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRAK1BP1
Entrez Gene ID
134728
Gene Name
interleukin 1 receptor associated kinase 1 binding protein 1
Gene Aliases
AIP70, SIMPL
UniGene
Hs.656212
Gene Chromosome Location
Chr.6: 78867486-78979411 on Build GRCh38
Transcripts
NM_001010844.3 XM_006715336.3 XM_011535447.1 XM_011535448.2 XM_017010264.1 XM_017010265.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7279584 c.167G>A p.S56N Chr.6: 78867743-78867743 on Build GRCh38
1187091 c.212T>A p.V71E Chr.6: 78867788-78867788 on Build GRCh38
321042 c.164T>C p.V55A Chr.6: 78867740-78867740 on Build GRCh38
1081834 c.149C>T p.T50I Chr.6: 78867725-78867725 on Build GRCh38
4625851 c.308G>A p.G103D Chr.6: 78867884-78867884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561101519 Chr.6: 78868150-78868150 on Build GRCh38
rs992765901 Chr.6: 78868033-78868033 on Build GRCh38
rs1034754547 Chr.6: 78868081-78868081 on Build GRCh38
rs770783743 Chr.6: 78867785-78867785 on Build GRCh38
rs760301172 Chr.6: 78867819-78867819 on Build GRCh38
rs9350789 Chr.6: 78867927-78867927 on Build GRCh38
rs948732183 Chr.6: 78867930-78867930 on Build GRCh38
rs760654285 Chr.6: 78867935-78867935 on Build GRCh38
rs185616488 Chr.6: 78868161-78868161 on Build GRCh38
rs934650211 Chr.6: 78868122-78868122 on Build GRCh38
rs9341753 Chr.6: 78868079-78868079 on Build GRCh38
rs866804003 Chr.6: 78867837-78867837 on Build GRCh38
rs781516528 Chr.6: 78867770-78867770 on Build GRCh38
rs865783471 Chr.6: 78868078-78868078 on Build GRCh38
rs545780584 Chr.6: 78867992-78867992 on Build GRCh38
rs1000693501 Chr.6: 78867957-78867957 on Build GRCh38
rs188400110 Chr.6: 78867969-78867969 on Build GRCh38
rs747764906 Chr.6: 78867770-78867771 on Build GRCh38
rs534885128 Chr.6: 78867859-78867859 on Build GRCh38
rs958714326 Chr.6: 78868031-78868031 on Build GRCh38
rs752775657 Chr.6: 78867938-78867938 on Build GRCh38
rs775563113 Chr.6: 78867807-78867807 on Build GRCh38
rs768722004 Chr.6: 78867769-78867769 on Build GRCh38
rs771961658 Chr.6: 78867885-78867885 on Build GRCh38
rs775934561 Chr.6: 78867915-78867915 on Build GRCh38
rs755438653 Chr.6: 78867929-78867929 on Build GRCh38
rs143611653 Chr.6: 78867847-78867847 on Build GRCh38
rs925933905 Chr.6: 78868113-78868113 on Build GRCh38
rs920389586 Chr.6: 78867824-78867824 on Build GRCh38
rs998006083 Chr.6: 78868019-78868019 on Build GRCh38
rs747153949 Chr.6: 78867768-78867768 on Build GRCh38
rs1040050640 Chr.6: 78868163-78868163 on Build GRCh38
rs765641986 Chr.6: 78867844-78867844 on Build GRCh38
rs922671758 Chr.6: 78868140-78868141 on Build GRCh38
rs775614101 Chr.6: 78867895-78867895 on Build GRCh38
rs772914982 Chr.6: 78867904-78867905 on Build GRCh38
rs989319555 Chr.6: 78868134-78868138 on Build GRCh38
rs193059889 Chr.6: 78868152-78868152 on Build GRCh38
rs61729435 Chr.6: 78867827-78867827 on Build GRCh38
rs1050132314 Chr.6: 78867832-78867832 on Build GRCh38
rs941238059 Chr.6: 78868165-78868165 on Build GRCh38
rs978756890 Chr.6: 78868112-78868112 on Build GRCh38
rs750075871 Chr.6: 78867922-78867922 on Build GRCh38
rs1043032939 Chr.6: 78867959-78867959 on Build GRCh38
rs921045591 Chr.6: 78867899-78867913 on Build GRCh38
rs556170745 Chr.6: 78868036-78868036 on Build GRCh38
rs1005254639 Chr.6: 78868040-78868040 on Build GRCh38
rs759222682 Chr.6: 78867800-78867800 on Build GRCh38
rs1052987034 Chr.6: 78867979-78867979 on Build GRCh38
rs1041337582 Chr.6: 78867862-78867862 on Build GRCh38
rs752083775 Chr.6: 78867928-78867928 on Build GRCh38
rs1023208525 Chr.6: 78868105-78868105 on Build GRCh38
rs745723861 Chr.6: 78867880-78867880 on Build GRCh38
rs1016606156 Chr.6: 78868053-78868053 on Build GRCh38
rs998497946 Chr.6: 78867775-78867775 on Build GRCh38
rs746277829 Chr.6: 78867753-78867754 on Build GRCh38
rs901459869 Chr.6: 78868183-78868183 on Build GRCh38
rs72901171 Chr.6: 78868153-78868153 on Build GRCh38
rs972295428 Chr.6: 78867881-78867881 on Build GRCh38
rs60805348 Chr.6: 78868088-78868088 on Build GRCh38
rs779600588 Chr.6: 78868189-78868189 on Build GRCh38
rs773903287 Chr.6: 78867769-78867770 on Build GRCh38
rs901015090 Chr.6: 78868005-78868005 on Build GRCh38
rs771494810 Chr.6: 78867828-78867829 on Build GRCh38
rs920424130 Chr.6: 78868095-78868095 on Build GRCh38
rs1024203923 Chr.6: 78868073-78868073 on Build GRCh38
rs1049675646 Chr.6: 78868020-78868020 on Build GRCh38
rs904645189 Chr.6: 78867956-78867956 on Build GRCh38
rs780609704 Chr.6: 78867850-78867850 on Build GRCh38
rs374362651 Chr.6: 78867838-78867838 on Build GRCh38
rs200563777 Chr.6: 78867767-78867767 on Build GRCh38
rs778042281 Chr.6: 78867879-78867879 on Build GRCh38
rs926080574 Chr.6: 78867964-78867965 on Build GRCh38
rs960488882 Chr.6: 78868085-78868085 on Build GRCh38
rs368333718 Chr.6: 78867911-78867911 on Build GRCh38
rs201035096 Chr.6: 78867852-78867852 on Build GRCh38
rs746414402 Chr.6: 78867760-78867760 on Build GRCh38
rs748435735 Chr.6: 78867868-78867868 on Build GRCh38
rs896924666 Chr.6: 78868071-78868073 on Build GRCh38
rs199667399 Chr.6: 78867813-78867813 on Build GRCh38
rs781085042 Chr.6: 78867863-78867863 on Build GRCh38
rs756754188 Chr.6: 78867997-78867997 on Build GRCh38
rs930459196 Chr.6: 78868130-78868130 on Build GRCh38
rs189265957 Chr.6: 78868214-78868214 on Build GRCh38
rs970282295 Chr.6: 78868107-78868107 on Build GRCh38
rs781338522 Chr.6: 78867933-78867933 on Build GRCh38
rs993508333 Chr.6: 78867916-78867916 on Build GRCh38
rs769848206 Chr.6: 78867878-78867878 on Build GRCh38
rs764859611 Chr.6: 78867921-78867921 on Build GRCh38
rs763671093 Chr.6: 78867820-78867820 on Build GRCh38
rs770143861 Chr.6: 78867765-78867765 on Build GRCh38
rs779096501 Chr.6: 78867744-78867744 on Build GRCh38
rs760638127 Chr.6: 78867905-78867905 on Build GRCh38
rs773979076 Chr.6: 78867786-78867786 on Build GRCh38

View Full Product Details