Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67293404-67293924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD19
Entrez Gene ID
146212
Gene Name
potassium channel tetramerization domain containing 19
Gene Aliases
-
UniGene
Hs.299127
Gene Chromosome Location
Chr.16: 67289490-67326758 on Build GRCh38
Transcripts
NM_001100915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5892289 c.1950G>A p.W650* Chr.16: 67293812-67293812 on Build GRCh38
4129190 c.1877A>C p.D626A Chr.16: 67293885-67293885 on Build GRCh38
225044 c.1867G>A p.E623K Chr.16: 67293895-67293895 on Build GRCh38
4774333 c.1905C>A p.L635L Chr.16: 67293857-67293857 on Build GRCh38
4129189 c.1881T>C p.P627P Chr.16: 67293881-67293881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776521902 Chr.16: 67293734-67293734 on Build GRCh38
rs745605880 Chr.16: 67293878-67293878 on Build GRCh38
rs921764183 Chr.16: 67293601-67293601 on Build GRCh38
rs757796226 Chr.16: 67293629-67293629 on Build GRCh38
rs756647138 Chr.16: 67293772-67293772 on Build GRCh38
rs780283089 Chr.16: 67293776-67293776 on Build GRCh38
rs752583700 Chr.16: 67293430-67293430 on Build GRCh38
rs768022868 Chr.16: 67293824-67293824 on Build GRCh38
rs777607123 Chr.16: 67293486-67293487 on Build GRCh38
rs564340076 Chr.16: 67293477-67293477 on Build GRCh38
rs758181872 Chr.16: 67293860-67293860 on Build GRCh38
rs748344817 Chr.16: 67293854-67293854 on Build GRCh38
rs774428720 Chr.16: 67293786-67293786 on Build GRCh38
rs766167431 Chr.16: 67293594-67293594 on Build GRCh38
rs1025342182 Chr.16: 67293619-67293619 on Build GRCh38
rs760662492 Chr.16: 67293574-67293574 on Build GRCh38
rs373585879 Chr.16: 67293836-67293836 on Build GRCh38
rs748583330 Chr.16: 67293502-67293502 on Build GRCh38
rs759631982 Chr.16: 67293540-67293540 on Build GRCh38
rs772244334 Chr.16: 67293867-67293867 on Build GRCh38
rs776776963 Chr.16: 67293530-67293530 on Build GRCh38
rs74901862 Chr.16: 67293885-67293885 on Build GRCh38
rs183031746 Chr.16: 67293843-67293843 on Build GRCh38
rs757003395 Chr.16: 67293876-67293876 on Build GRCh38
rs778195475 Chr.16: 67293528-67293528 on Build GRCh38
rs745983571 Chr.16: 67293632-67293632 on Build GRCh38
rs147536831 Chr.16: 67293558-67293558 on Build GRCh38
rs754484386 Chr.16: 67293611-67293611 on Build GRCh38
rs776220578 Chr.16: 67293437-67293437 on Build GRCh38
rs145649592 Chr.16: 67293739-67293739 on Build GRCh38
rs757942684 Chr.16: 67293618-67293618 on Build GRCh38
rs779890887 Chr.16: 67293877-67293878 on Build GRCh38
rs762572868 Chr.16: 67293882-67293882 on Build GRCh38
rs780690255 Chr.16: 67293839-67293839 on Build GRCh38
rs374733007 Chr.16: 67293740-67293740 on Build GRCh38
rs931365634 Chr.16: 67293441-67293441 on Build GRCh38
rs374684640 Chr.16: 67293454-67293454 on Build GRCh38
rs769971827 Chr.16: 67293633-67293633 on Build GRCh38
rs747271819 Chr.16: 67293627-67293627 on Build GRCh38
rs749555072 Chr.16: 67293680-67293680 on Build GRCh38
rs140145155 Chr.16: 67293700-67293700 on Build GRCh38
rs757286144 Chr.16: 67293596-67293607 on Build GRCh38
rs778232585 Chr.16: 67293625-67293625 on Build GRCh38
rs572214166 Chr.16: 67293437-67293437 on Build GRCh38
rs781776908 Chr.16: 67293767-67293767 on Build GRCh38
rs750748982 Chr.16: 67293832-67293832 on Build GRCh38
rs369962992 Chr.16: 67293789-67293789 on Build GRCh38
rs748689213 Chr.16: 67293484-67293484 on Build GRCh38
rs552329466 Chr.16: 67293770-67293770 on Build GRCh38
rs747575214 Chr.16: 67293529-67293529 on Build GRCh38
rs760369250 Chr.16: 67293723-67293723 on Build GRCh38
rs996165480 Chr.16: 67293443-67293443 on Build GRCh38
rs567420800 Chr.16: 67293641-67293642 on Build GRCh38
rs371352280 Chr.16: 67293782-67293782 on Build GRCh38
rs750873550 Chr.16: 67293891-67293891 on Build GRCh38
rs201863465 Chr.16: 67293701-67293701 on Build GRCh38
rs1044560998 Chr.16: 67293442-67293442 on Build GRCh38
rs759100184 Chr.16: 67293735-67293735 on Build GRCh38
rs767217223 Chr.16: 67293889-67293889 on Build GRCh38
rs200929259 Chr.16: 67293787-67293787 on Build GRCh38
rs779247568 Chr.16: 67293845-67293845 on Build GRCh38
rs559952169 Chr.16: 67293745-67293745 on Build GRCh38
rs201293471 Chr.16: 67293761-67293761 on Build GRCh38
rs1042046722 Chr.16: 67293496-67293496 on Build GRCh38
rs754978796 Chr.16: 67293844-67293844 on Build GRCh38
rs763873691 Chr.16: 67293811-67293811 on Build GRCh38
rs751149420 Chr.16: 67293814-67293814 on Build GRCh38
rs374291048 Chr.16: 67293522-67293522 on Build GRCh38
rs745792974 Chr.16: 67293654-67293655 on Build GRCh38
rs199518783 Chr.16: 67293880-67293880 on Build GRCh38
rs190654124 Chr.16: 67293448-67293448 on Build GRCh38
rs950400045 Chr.16: 67293459-67293459 on Build GRCh38
rs773869253 Chr.16: 67293886-67293886 on Build GRCh38
rs919901274 Chr.16: 67293682-67293682 on Build GRCh38
rs974493741 Chr.16: 67293599-67293599 on Build GRCh38
rs570733590 Chr.16: 67293703-67293703 on Build GRCh38
rs1044069772 Chr.16: 67293692-67293692 on Build GRCh38
rs77164997 Chr.16: 67293817-67293817 on Build GRCh38
rs781761430 Chr.16: 67293631-67293631 on Build GRCh38
rs983096489 Chr.16: 67293460-67293460 on Build GRCh38
rs762357193 Chr.16: 67293807-67293807 on Build GRCh38
rs202125861 Chr.16: 67293606-67293606 on Build GRCh38
rs371252010 Chr.16: 67293598-67293598 on Build GRCh38
rs769946132 Chr.16: 67293885-67293888 on Build GRCh38
rs927444554 Chr.16: 67293447-67293447 on Build GRCh38
rs770207839 Chr.16: 67293554-67293554 on Build GRCh38
rs772840477 Chr.16: 67293572-67293572 on Build GRCh38
rs970630593 Chr.16: 67293611-67293612 on Build GRCh38
rs200302400 Chr.16: 67293881-67293881 on Build GRCh38
rs533042540 Chr.16: 67293717-67293717 on Build GRCh38
rs773069676 Chr.16: 67293708-67293708 on Build GRCh38
rs73592842 Chr.16: 67293567-67293567 on Build GRCh38
rs372339467 Chr.16: 67293642-67293642 on Build GRCh38
rs753343039 Chr.16: 67293504-67293504 on Build GRCh38
rs529630236 Chr.16: 67293724-67293724 on Build GRCh38
rs757530064 Chr.16: 67293765-67293765 on Build GRCh38
rs778025965 Chr.16: 67293436-67293436 on Build GRCh38
rs1045118742 Chr.16: 67293853-67293853 on Build GRCh38
rs143567769 Chr.16: 67293641-67293641 on Build GRCh38
rs780973499 Chr.16: 67293877-67293877 on Build GRCh38
rs972719458 Chr.16: 67293672-67293672 on Build GRCh38
rs375929826 Chr.16: 67293614-67293614 on Build GRCh38
rs748374625 Chr.16: 67293885-67293889 on Build GRCh38
rs763596193 Chr.16: 67293752-67293752 on Build GRCh38
rs570644111 Chr.16: 67293777-67293777 on Build GRCh38
rs1009224847 Chr.16: 67293486-67293486 on Build GRCh38
rs769654789 Chr.16: 67293802-67293802 on Build GRCh38
rs200456334 Chr.16: 67293822-67293822 on Build GRCh38

View Full Product Details