Gene Transcripts

Gene Symbol
SDK2
Entrez Gene ID
54549
Gene Name
sidekick cell adhesion molecule 2
Gene Aliases
Chromosome Location
Chr. 17: 73334384 - 73644482 on Build GRCh38
UniGene ID
Hs.435719
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144952.1 NP_001138424.1 45 7195
XM_011524914.2 XP_011523216.1 44 7522
XM_011524915.2 XP_011523217.1 46 7252
GenBank mRNA BC066363.1 - 37 6095

View Full Product Details