Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 83273522-83274017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC90B
Entrez Gene ID
60492
Gene Name
coiled-coil domain containing 90B
Gene Aliases
MDS011, MDS025
UniGene
Hs.368866
Gene Chromosome Location
Chr.11: 83259093-83286407 on Build GRCh38
Transcripts
NM_001286116.1 NM_001286117.1 NM_001286118.1 NM_001286119.1 NM_001286120.1 NM_021825.4 XM_005274154.2 XM_005274156.3 XM_005274159.3 XM_017018103.1 XM_017018104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6846114 c.403A>G p.M135V Chr.11: 83273989-83273989 on Build GRCh38
5505324 c.441+9A>T p.? Chr.11: 83273942-83273942 on Build GRCh38
1746610 c.521G>A p.R174K Chr.11: 83273812-83273812 on Build GRCh38
3810550 c.582A>T p.E194D Chr.11: 83273659-83273659 on Build GRCh38
5505323 c.468+9A>T p.? Chr.11: 83273942-83273942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758806973 Chr.11: 83273599-83273599 on Build GRCh38
rs763439647 Chr.11: 83273695-83273695 on Build GRCh38
rs376963220 Chr.11: 83273703-83273703 on Build GRCh38
rs768501400 Chr.11: 83273661-83273661 on Build GRCh38
rs748976429 Chr.11: 83273660-83273660 on Build GRCh38
rs781399502 Chr.11: 83273628-83273628 on Build GRCh38
rs777793696 Chr.11: 83273608-83273608 on Build GRCh38
rs772674254 Chr.11: 83273678-83273678 on Build GRCh38
rs771263245 Chr.11: 83273933-83273934 on Build GRCh38
rs762388859 Chr.11: 83273812-83273812 on Build GRCh38
rs201678091 Chr.11: 83273929-83273929 on Build GRCh38
rs748276000 Chr.11: 83273793-83273795 on Build GRCh38
rs1041566500 Chr.11: 83273648-83273648 on Build GRCh38
rs774737868 Chr.11: 83273827-83273827 on Build GRCh38
rs764945950 Chr.11: 83273677-83273677 on Build GRCh38
rs60237170 Chr.11: 83273736-83273737 on Build GRCh38
rs746910132 Chr.11: 83273728-83273728 on Build GRCh38
rs756274892 Chr.11: 83273718-83273718 on Build GRCh38
rs758199474 Chr.11: 83273939-83273939 on Build GRCh38
rs749243705 Chr.11: 83273759-83273759 on Build GRCh38
rs767883740 Chr.11: 83273706-83273706 on Build GRCh38
rs977290269 Chr.11: 83273917-83273917 on Build GRCh38
rs776072895 Chr.11: 83273694-83273694 on Build GRCh38
rs557547702 Chr.11: 83273635-83273635 on Build GRCh38
rs764549560 Chr.11: 83273825-83273825 on Build GRCh38
rs543562 Chr.11: 83273942-83273942 on Build GRCh38
rs983568218 Chr.11: 83273870-83273870 on Build GRCh38
rs763498330 Chr.11: 83273811-83273811 on Build GRCh38
rs992466944 Chr.11: 83273900-83273900 on Build GRCh38
rs973642731 Chr.11: 83273582-83273582 on Build GRCh38
rs750677417 Chr.11: 83273856-83273856 on Build GRCh38
rs904351541 Chr.11: 83273674-83273674 on Build GRCh38
rs757482484 Chr.11: 83273624-83273624 on Build GRCh38
rs778743571 Chr.11: 83273765-83273765 on Build GRCh38
rs746653817 Chr.11: 83273945-83273945 on Build GRCh38
rs751921310 Chr.11: 83273698-83273698 on Build GRCh38
rs945873077 Chr.11: 83273629-83273629 on Build GRCh38
rs772729595 Chr.11: 83273954-83273954 on Build GRCh38
rs780068167 Chr.11: 83273748-83273748 on Build GRCh38
rs745402250 Chr.11: 83273974-83273974 on Build GRCh38
rs772870702 Chr.11: 83273794-83273794 on Build GRCh38
rs899783273 Chr.11: 83273809-83273809 on Build GRCh38
rs776178985 Chr.11: 83273957-83273957 on Build GRCh38
rs749606068 Chr.11: 83273931-83273931 on Build GRCh38
rs774081110 Chr.11: 83273669-83273669 on Build GRCh38
rs368936447 Chr.11: 83273923-83273923 on Build GRCh38
rs563753212 Chr.11: 83273906-83273906 on Build GRCh38
rs372262391 Chr.11: 83273760-83273760 on Build GRCh38
rs541952587 Chr.11: 83273851-83273851 on Build GRCh38
rs537068438 Chr.11: 83273710-83273710 on Build GRCh38
rs780164577 Chr.11: 83273652-83273652 on Build GRCh38
rs769123058 Chr.11: 83273746-83273747 on Build GRCh38
rs1019615614 Chr.11: 83273737-83273737 on Build GRCh38
rs781121858 Chr.11: 83273754-83273757 on Build GRCh38
rs200843474 Chr.11: 83273597-83273597 on Build GRCh38
rs769157843 Chr.11: 83273987-83273987 on Build GRCh38
rs146002262 Chr.11: 83273727-83273728 on Build GRCh38
rs548957417 Chr.11: 83273702-83273703 on Build GRCh38
rs555971139 Chr.11: 83273830-83273830 on Build GRCh38
rs760034134 Chr.11: 83273964-83273964 on Build GRCh38
rs746453343 Chr.11: 83273797-83273797 on Build GRCh38
rs747930280 Chr.11: 83273775-83273775 on Build GRCh38
rs751805936 Chr.11: 83273620-83273620 on Build GRCh38
rs755028857 Chr.11: 83273728-83273730 on Build GRCh38
rs376807996 Chr.11: 83273910-83273910 on Build GRCh38
rs1030353364 Chr.11: 83273852-83273852 on Build GRCh38
rs146528114 Chr.11: 83273684-83273684 on Build GRCh38
rs760085416 Chr.11: 83273618-83273618 on Build GRCh38
rs758289827 Chr.11: 83273721-83273721 on Build GRCh38
rs762292538 Chr.11: 83273845-83273845 on Build GRCh38
rs376200701 Chr.11: 83273683-83273683 on Build GRCh38
rs994150554 Chr.11: 83273777-83273777 on Build GRCh38
rs190805303 Chr.11: 83273670-83273670 on Build GRCh38
rs547351855 Chr.11: 83273994-83273994 on Build GRCh38
rs916893353 Chr.11: 83273908-83273908 on Build GRCh38
rs766469343 Chr.11: 83273905-83273905 on Build GRCh38
rs113915821 Chr.11: 83273943-83273943 on Build GRCh38
rs756469287 Chr.11: 83273882-83273882 on Build GRCh38
rs771669979 Chr.11: 83273784-83273784 on Build GRCh38
rs140971996 Chr.11: 83273804-83273804 on Build GRCh38
rs942174723 Chr.11: 83273549-83273549 on Build GRCh38
rs773517708 Chr.11: 83273848-83273861 on Build GRCh38
rs1034072970 Chr.11: 83273829-83273829 on Build GRCh38
rs375738671 Chr.11: 83273949-83273949 on Build GRCh38
rs993168099 Chr.11: 83273571-83273571 on Build GRCh38
rs763566659 Chr.11: 83273627-83273628 on Build GRCh38
rs756800822 Chr.11: 83273669-83273669 on Build GRCh38
rs778743119 Chr.11: 83273914-83273914 on Build GRCh38
rs369921990 Chr.11: 83273704-83273704 on Build GRCh38
rs954858157 Chr.11: 83273869-83273869 on Build GRCh38
rs753456737 Chr.11: 83273648-83273648 on Build GRCh38
rs973506928 Chr.11: 83273933-83273933 on Build GRCh38
rs769845880 Chr.11: 83273812-83273815 on Build GRCh38
rs182162743 Chr.11: 83273675-83273675 on Build GRCh38
rs774739471 Chr.11: 83273934-83273937 on Build GRCh38
rs776067402 Chr.11: 83273810-83273810 on Build GRCh38

View Full Product Details