Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3750024-3750289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA12B
Entrez Gene ID
116835
Gene Name
heat shock protein family A (Hsp70) member 12B
Gene Aliases
C20orf60
UniGene
Hs.516854
Gene Chromosome Location
Chr.20: 3732670-3753111 on Build GRCh38
Transcripts
NM_001197327.1 NM_001318322.1 NM_052970.4 XM_011529151.2 XM_017027632.1 XM_017027633.1 XM_017027634.1 XM_017027635.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4645396 c.1215G>A p.A405A Chr.20: 3750141-3750141 on Build GRCh38
6682553 c.1181G>A p.R394H Chr.20: 3750107-3750107 on Build GRCh38
5517235 c.1227G>A p.S409S Chr.20: 3750153-3750153 on Build GRCh38
1411715 c.1174G>A p.E392K Chr.20: 3750100-3750100 on Build GRCh38
3963554 c.1249T>A p.F417I Chr.20: 3750175-3750175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770254202 Chr.20: 3750269-3750269 on Build GRCh38
rs370598013 Chr.20: 3750221-3750221 on Build GRCh38
rs771173094 Chr.20: 3750267-3750267 on Build GRCh38
rs756031970 Chr.20: 3750134-3750134 on Build GRCh38
rs763765071 Chr.20: 3750211-3750211 on Build GRCh38
rs751082743 Chr.20: 3750215-3750215 on Build GRCh38
rs141564153 Chr.20: 3750185-3750185 on Build GRCh38
rs755874590 Chr.20: 3750227-3750227 on Build GRCh38
rs73895608 Chr.20: 3750257-3750257 on Build GRCh38
rs748187474 Chr.20: 3750140-3750140 on Build GRCh38
rs139310351 Chr.20: 3750137-3750137 on Build GRCh38
rs372363417 Chr.20: 3750136-3750136 on Build GRCh38
rs944947114 Chr.20: 3750066-3750066 on Build GRCh38
rs771119929 Chr.20: 3750169-3750169 on Build GRCh38
rs781177119 Chr.20: 3750132-3750132 on Build GRCh38
rs144840007 Chr.20: 3750122-3750122 on Build GRCh38
rs747041765 Chr.20: 3750264-3750264 on Build GRCh38
rs375725643 Chr.20: 3750087-3750087 on Build GRCh38
rs367586620 Chr.20: 3750058-3750058 on Build GRCh38
rs765725768 Chr.20: 3750093-3750093 on Build GRCh38
rs369126307 Chr.20: 3750268-3750268 on Build GRCh38
rs979776341 Chr.20: 3750202-3750202 on Build GRCh38
rs747268390 Chr.20: 3750162-3750162 on Build GRCh38
rs766795451 Chr.20: 3750084-3750084 on Build GRCh38
rs757023154 Chr.20: 3750220-3750220 on Build GRCh38
rs368513665 Chr.20: 3750120-3750120 on Build GRCh38
rs745657847 Chr.20: 3750133-3750133 on Build GRCh38
rs971077838 Chr.20: 3750193-3750193 on Build GRCh38
rs760872422 Chr.20: 3750073-3750073 on Build GRCh38
rs111540710 Chr.20: 3750069-3750069 on Build GRCh38
rs754875285 Chr.20: 3750248-3750248 on Build GRCh38
rs1005081550 Chr.20: 3750177-3750177 on Build GRCh38
rs570002442 Chr.20: 3750222-3750222 on Build GRCh38
rs773362896 Chr.20: 3750068-3750068 on Build GRCh38
rs372853784 Chr.20: 3750063-3750063 on Build GRCh38
rs1054810962 Chr.20: 3750156-3750156 on Build GRCh38
rs957276054 Chr.20: 3750223-3750223 on Build GRCh38
rs757855596 Chr.20: 3750113-3750113 on Build GRCh38
rs200973186 Chr.20: 3750231-3750231 on Build GRCh38
rs776009719 Chr.20: 3750201-3750201 on Build GRCh38
rs752178720 Chr.20: 3750111-3750111 on Build GRCh38
rs773272336 Chr.20: 3750153-3750153 on Build GRCh38
rs760009470 Chr.20: 3750090-3750090 on Build GRCh38
rs530230071 Chr.20: 3750179-3750179 on Build GRCh38
rs759904018 Chr.20: 3750184-3750184 on Build GRCh38
rs199719933 Chr.20: 3750054-3750054 on Build GRCh38
rs763853731 Chr.20: 3750114-3750114 on Build GRCh38
rs753876891 Chr.20: 3750245-3750245 on Build GRCh38
rs763542746 Chr.20: 3750210-3750210 on Build GRCh38

View Full Product Details