Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252520.1 | NP_001239449.1 | 4 | 725 | |
NM_001252521.1 | NP_001239450.1 | 4 | 725 | ||
NM_001252522.1 | NP_001239451.1 | 4 | 725 | ||
NM_001252523.1 | NP_001239452.1 | 4 | 725 | ||
NM_001252524.1 | NP_001239453.1 | 4 | 725 | ||
NM_146194.4 | NP_666306.2 | 4 | 725 | ||
GenBank mRNA | AK144960.1 | - | 4 | 463 | |
AK162360.1 | - | 4 | 418 | ||
AK170373.1 | - | 4 | 745 | ||
AK220291.1 | - | 4 | 441 | ||
AY206701.1 | - | 4 | 409 | ||
BC011470.1 | - | 4 | 435 | 2651109 | |
BC021491.1 | - | 4 | 674 | 5345558 | |
BC023843.1 | - | 4 | 473 | 5346041 | |
BC025566.1 | - | 4 | 748 | 5322606 | |
BC027116.1 | - | 1 | 36 | ||
BC057683.1 | - | 4 | 676 | 3671962 |