Gene Transcripts

Gene Symbol
NCSTN
Entrez Gene ID
23385
Gene Name
nicastrin
Gene Aliases
ATAG1874
Chromosome Location
Chr. 1: 160343056 - 160358952 on Build GRCh38
UniGene ID
Hs.517249
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290184.1 NP_001277113.1 17 2247
NM_001290186.1 NP_001277115.1 13 1694
NM_015331.2 NP_056146.1 16 2108
GenBank mRNA AF240468.1 - 16 2126
AK296153.1 - 11 1393
AK299142.1 - 13 1581
AK314764.1 - 16 2015
AY359120.1 - 16 1985
BC047621.1 - 17 2204 4827966
D87442.1 - 16 1982

View Full Product Details