Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001242466.1 | NP_001229395.1 | ||||
NM_181504.3 | NP_852556.2 | |||||
NM_181523.2 | NP_852664.1 | |||||
NM_181524.1 | NP_852665.1 | |||||
XM_005248542.3 | XP_005248599.1 | |||||
XM_011543493.2 | XP_011541795.1 | |||||
XM_017009585.1 | XP_016865074.1 | |||||
XM_017009586.1 | XP_016865075.1 | |||||
GenBank mRNA | AF279367.1 | - | ||||
AI124626.1 | - | |||||
AI334281.1 | - | |||||
AK000121.1 | - | |||||
AK094785.1 | - | |||||
AK126345.1 | - | |||||
AK223613.1 | - | |||||
AK294919.1 | - | |||||
AK299743.1 | - | |||||
AW630519.1 | - | |||||
BC030815.1 | - | 4290954 | ||||
BC094795.1 | - | 30528412 | ||||
BE888150.1 | - | |||||
BQ723333.1 | - | |||||
CA427864.1 | - | |||||
CB216668.1 | - | |||||
CT003423.1 | - | |||||
DA487709.1 | - | |||||
ENST00000521381.0 | - | |||||
JX133164.1 | - | |||||
JX133165.1 | - | |||||
M61906.1 | - | |||||
U49349.1 | - |