Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_020529.2 | NP_065390.1 | ||||
GenBank mRNA | AK311515.1 | - | ||||
AK313421.1 | - | |||||
AY033600.1 | - | |||||
BC002601.2 | - | 3161686 | ||||
BC004983.1 | - | 2957970 | ||||
BT007091.1 | - | |||||
BU737609.1 | - | |||||
M69043.1 | - |