Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001207012.1 | NP_001193941.1 | ||||
NM_001293643.1 | NP_001280572.1 | |||||
NM_002632.5 | NP_002623.2 | |||||
GenBank mRNA | A18411.1 | - | ||||
AK023843.1 | - | |||||
AK309886.1 | - | |||||
AK310608.1 | - | |||||
AK310886.1 | - | |||||
AL543693.3 | - | |||||
BC001422.2 | - | 3139175 | ||||
BC007255.1 | - | 3028008 | ||||
BC007789.2 | - | 3629125 | ||||
BT007182.1 | - | |||||
BX248289.1 | - | |||||
BX356823.2 | - | |||||
ENST00000555567.0 | - | |||||
S72960.1 | - | |||||
X54936.1 | - |