Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFBI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000358.2 | NP_000349.1 | ||||
GenBank mRNA | AB209598.1 | - | ||||
AK093916.1 | - | |||||
AK094055.1 | - | |||||
AK094581.1 | - | |||||
AK222833.1 | - | |||||
AK298603.1 | - | |||||
AK303643.1 | - | |||||
BC000097.2 | - | 3506051 | ||||
BC004972.1 | - | 2958878 | ||||
BC026352.1 | - | |||||
BT009820.1 | - | |||||
DA051240.1 | - | |||||
ENST00000442011.0 | - | |||||
KU178357.1 | - | |||||
KU178358.1 | - | |||||
M77349.1 | - |