Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001024955.2 | NP_001020126.1 | ||||
NM_001077495.2 | NP_001070963.1 | |||||
GenBank mRNA | AI838690.1 | - | ||||
AK046259.1 | - | |||||
AK049798.1 | - | |||||
AK138868.1 | - | |||||
AK160493.1 | - | |||||
AK164783.1 | - | |||||
AK171366.1 | - | |||||
BB647657.1 | - | |||||
BC002168.1 | - | |||||
BC026146.1 | - | 3979333 | ||||
BC051106.1 | - | 5031261 | ||||
BM963795.1 | - | |||||
BY276314.1 | - | |||||
CB519962.1 | - | |||||
CF169061.1 | - | |||||
M60651.1 | - | |||||
U50413.1 | - | |||||
U50414.1 | - |