Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001252600.1 | NP_001239529.1 | ||||
NM_001252601.1 | NP_001239530.1 | |||||
NM_016850.3 | NP_058546.1 | |||||
GenBank mRNA | AK002830.1 | - | ||||
AK004349.1 | - | |||||
AK079685.1 | - | |||||
AK089869.1 | - | |||||
AK150403.1 | - | |||||
AK150608.1 | - | |||||
AK151470.1 | - | |||||
AK152401.1 | - | |||||
AK152403.1 | - | |||||
AK153054.1 | - | |||||
AK155950.1 | - | |||||
AK158378.1 | - | |||||
AK158403.1 | - | |||||
AK159142.1 | - | |||||
AK159883.1 | - | |||||
BC138799.1 | - | 8861821 | ||||
BC145422.1 | - | |||||
BQ033099.1 | - | |||||
BY155234.1 | - | |||||
DT923384.1 | - | |||||
U73037.1 | - |