Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_029094.3 | NP_083370.2 | ||||
GenBank mRNA | AK003230.1 | - | ||||
AK081350.1 | - | |||||
AK090116.1 | - | |||||
AK154106.1 | - | |||||
AK165230.1 | - | |||||
BC039650.1 | - | |||||
BC127073.1 | - | 40110218 | ||||
CJ129150.1 | - | |||||
CV558291.1 | - |